2-Bromopyridine ≥95%

Supplier: Matrix Scientific

074988-100G 074988-25G
102539-198EA 204.62 USD
102539-198 102833-752
2-Bromopyridine ≥95%
2-Bromopyridine

Hazard: Toxic

Formula: C₅H₄BrN
MW: 158 g/mol
Boiling Pt: 192
Melting Pt: -40.1
Density: 1.625
Flash Pt: 97 °C
MDL Number: MFCD00006219
CAS Number: 109-04-6
UN: 2810
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR