4-Ethynylbenzoic acid ≥95%

Supplier: Matrix Scientific

074986-1G 074986-5G
102539-186EA 89.23 USD
102539-186 102539-188
4-Ethynylbenzoic acid ≥95%
4-Ethynylbenzoic acid

Hazard: Irritant

Formula: C₉H₆O₂
MDL Number: MFCD00168819
CAS Number: 10602-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR