3,5-Pyrazoledicarboxylic acid ≥95%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
Pyrazole-3,5-dicarboxylic acid, 1H-Pyrazole-3,5-dicarboxylic acid
074984-25G
074984-100G
102551-932EA
289.23
USD
102551-932
102551-934
3,5-Pyrazoledicarboxylic acid ≥95%
3,5-Pyrazoledicarboxylic acid
Purity: 95+%
CAS: 3112-31-0
MDL Number: MFCD00005235
Molecular Formula: C5H4N2O4
Molecular Weight: 156,1
Hazard: Irritant
Formula:
C₅H₄N₂O₄ MW: 156.10 g/mol Melting Pt: 287 °C |
MDL Number:
MFCD00005235 CAS Number: 3112-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...