4,5-Dicyanoimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 4,5-Dicyano-1H-imidazole, Imidazole-4,5-dicarbonitrile

074981-5G 074981-10G 074981-100G
102539-158EA 72.31 USD
102539-158 102539-160 102539-162
4,5-Dicyanoimidazole ≥95%
4,5-Dicyanoimidazole

Hazard: Irritant

Formula: C₅H₂N₄
MW: 118.1 g/mol
MDL Number: MFCD00005194
CAS Number: 1122-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR