DL-Glutamine ≥95%

Supplier: Matrix Scientific

Synonyms: H-DL-Gln-OH, 2,5-Diamino-5-oxopentanoic acid

074979-10G 074979-100G
102539-148EA 53.85 USD
102539-148 102539-150
DL-Glutamine ≥95%
DL-Glutamine

Hazard: Irritant

Formula: C₅H₁₀N₂O₃
MW: 146.144 g/mol
Melting Pt: 186 °C
MDL Number: MFCD00065103
CAS Number: 6899-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR