(±)-2-Piperazinecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Piperazine-2-carboxylic acid

074978-5G 074978-10G 074978-100G
102539-142EA 107.69 USD
102539-142 102539-144 102539-146
(±)-2-Piperazinecarboxylic acid ≥95%
(±)-2-Piperazinecarboxylic acid

Hazard: Irritant

Formula: C₅H₁₀N₂O₂
MDL Number: MFCD00132878
CAS Number: 2762-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR