2-Chloro-4-nitrobenzoic acid ≥95%
Supplier: Matrix Scientific
074975-10G
074975-100G
102539-128EA
36.92
USD
102539-128
102539-130
2-Chloro-4-nitrobenzoic acid ≥95%
2-Chloro-4-nitrobenzoic acid
Hazard: Irritant
Formula:
C₇H₄ClNO₄ MW: 201.57 g/mol |
MDL Number:
MFCD00007209 CAS Number: 99-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...