5-Nitro-o-toluidine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Nitro-2-methylaniline, 2-Methyl-5-nitroaniline, 2-Amino-4-nitrotoluene

074974-100G 074974-500G
102539-126EA 49.23 USD
102539-126 102551-930
5-Nitro-o-toluidine ≥95%
5-Nitro-o-toluidine

Hazard: Toxic

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 108
MDL Number: MFCD00007741
CAS Number: 99-55-8
UN: 2660
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR