3,5-Dinitrobenzoic acid ≥95%

Supplier: Matrix Scientific

074973-10G 074973-100G
102539-120EA 36.92 USD
102539-120 102539-122
3,5-Dinitrobenzoic acid ≥95%
3,5-Dinitrobenzoic acid

Hazard: Irritant

Formula: C₇H₄N₂O₆
Melting Pt: 204…208 °C
MDL Number: MFCD00007253
CAS Number: 99-34-3
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR