H-β-HoPhe-OH.HCl ≥95%

Supplier: Matrix Scientific

Synonyms: L-β-Homophenylalanine hydrochloride, (S)-3-Amino-4-phenylbutyric acid hydrochloride

074969-1G 074969-5G
102539-102EA 640 USD
102539-102 102539-104
H-β-HoPhe-OH.HCl ≥95%
H-β-HoPhe-OH.HCl

Hazard: Irritant

Formula: C₁₀H₁₄ClNO₂
MW: 215.67 g/mol
MDL Number: MFCD01862875
CAS Number: 138165-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR