2-Nitro-p-toluidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-2-nitroaniline, 4-Amino-3-nitrotoluene, 2-Nitro-4-methylaniline

074965-10G 074965-100G
102539-082EA 36.92 USD
102539-082 102539-084
2-Nitro-p-toluidine ≥95%
2-Nitro-p-toluidine

Hazard: Irritant

Formula: C₇H₈N₂O₂
MDL Number: MFCD00007907
CAS Number: 89-62-3
UN: 2660
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR