2-(Chloromethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

074961-1G 074961-5G 074961-10G
102539-062EA 100 USD
102539-062 102539-064 102539-066
2-(Chloromethyl)benzoic acid ≥95%
2-(Chloromethyl)benzoic acid

Hazard: Irritant

Formula: C₈H₇ClO₂
MDL Number: MFCD06208434
CAS Number: 85888-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR