trans-4-Dimethylaminocrotonic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (E)-4-Dimethylaminocrotonic acid hydrochloride, (E)-4-(Dimethylamino)-2-butenoic acid hydrochloride, (E)-4-(Dimethylamino)but-2-enoic acid hydrochloride

074960-1G 074960-5G
102539-058EA 60 USD
102539-058 102539-060
trans-4-Dimethylaminocrotonic acid hydrochloride ≥95%
trans-4-Dimethylaminocrotonic acid hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO₂
MW: 165.62 g/mol
MDL Number: MFCD03695466
CAS Number: 848133-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR