N-Boc-D-prolinol ≥95%
Supplier: Matrix Scientific
Synonyms:
(R)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (R)-1-Boc-2-pyrrolidinemethanol, (R)-(+)-1-Boc-2-pyrrolidinemethanol, (R)-(+)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, N-tert-Butoxycarbonyl-D-prolinol, N-t-Boc-D-prolinol, Boc-D-Prolinol
074959-1G
074959-5G
074959-10G
102539-052EA
32.31
USD
102539-052
102539-054
102539-056
N-Boc-D-prolinol ≥95%
N-Boc-D-prolinol
Hazard: Irritant
Formula:
C₁₀H₁₉NO₃ MW: 201.3 g/mol |
MDL Number:
MFCD00040580 CAS Number: 83435-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...