2-Cyanobenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Formylbenzonitrile

074953-1G 074953-5G
102539-020EA 290.77 USD
102539-020 102539-022
2-Cyanobenzaldehyde ≥95%
2-Cyanobenzaldehyde

Hazard: Irritant

Formula: C₈H₅NO
MW: 131.13 g/mol
MDL Number: MFCD00017503
CAS Number: 7468-67-9
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR