(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥95%

Supplier: Matrix Scientific

Synonyms: D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-​(+)​-​Phenyllactic acid

074952-1G 074952-5G 074952-25G
102539-014EA 56.92 USD
102539-014 102539-016 102664-020
(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥95%
(R)-(+)-2-Hydroxy-3-phenylpropionic acid

Hazard: Irritant

Formula: C₉H₁₀O₃
MW: 166.1739 g/mol
MDL Number: MFCD00078062
CAS Number: 7326-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR