Boc-Glu-OMe ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-4-((tert-Butoxycarbonyl)amino)-5-methoxy-5-oxopentanoic acid, Boc-Glu-OMe

074950-5G 074950-25G
102539-006EA 176.92 USD
102539-006 102551-926
Boc-Glu-OMe ≥95%
Boc-Glu-OMe

Hazard: Irritant

Formula: C₁₁H₁₉NO₆
MW: 261.2717 g/mol
MDL Number: MFCD00076931
CAS Number: 72086-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR