DL-ɑ-Amino-ε-caprolactam ≥95%

Supplier: Matrix Scientific

Synonyms: DL-3-Amino-2-oxohexamethyleneimine, DL-3-Aminohexahydro-2-azepinone, 3-Amino-2-azepanone(Chunks or pellets or lump), 3-Amino-2-azepanone

074944-1G 074944-5G
102538-976EA 92.31 USD
102538-976 102538-978
DL-ɑ-Amino-ε-caprolactam ≥95%
DL-alpha-Amino epsilon caprolactam

Hazard: Irritant

Formula: C₆H₁₂N₂O
MW: 128.18 g/mol
Boiling Pt: 168-172°C/13 Torr
Melting Pt: 68…69 °C
MDL Number: MFCD00064475
CAS Number: 671-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR