4-Chlorobutyraldehyde diethyl acetal ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-1,1-diethoxybutane, 4-Chlorobutanal diethyl acetal

074938-1G 074938-5G
102538-948EA 193.85 USD
102538-948 102538-950
4-Chlorobutyraldehyde diethyl acetal ≥95%
4-Chlorobutyraldehyde diethyl acetal

Hazard: Irritant

Formula: C₈H₁₇ClO₂
MDL Number: MFCD00671476
CAS Number: 6139-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR