2,3,4-Trimethoxybenzoic acid ≥95%

Supplier: Matrix Scientific

074937-10G 074937-100G
102538-944EA 72.31 USD
102538-944 102538-946
2,3,4-Trimethoxybenzoic acid ≥95%
2,3,4-Trimethoxybenzoic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₅
MW: 212.2 g/mol
MDL Number: MFCD00002433
CAS Number: 573-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR