Benzyltriethylammonium chloride ≥95%

Supplier: Matrix Scientific

074931-10G 074931-100G
102538-920EA 30.77 USD
102538-920 102538-922
Benzyltriethylammonium chloride ≥95%
Benzyltriethylammonium chloride

Hazard: Irritant

Formula: C₁₃H₂₂ClN
MDL Number: MFCD00011824
CAS Number: 56-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR