4-Aminobutyric acid ≥95%

Supplier: Matrix Scientific

074930-10G 074930-100G
102538-916EA 36.92 USD
102538-916 102538-918
4-Aminobutyric acid ≥95%
4-Aminobutyric acid

Hazard: Irritant

Formula: C₄H₉NO₂
MDL Number: MFCD00008226
CAS Number: 56-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR