D-(-)-Leucinol ≥95%

Supplier: Matrix Scientific

Synonyms: D-Leucinol, (R)-(-)-Leucinol, (R)-(-)-2-Amino-4-methylpentan-1-ol, (R)-2-Amino-4-methylpentan-1-ol

074925-1G 074925-5G 074925-10G
102538-894EA 35.38 USD
102538-894 102538-896 102538-898
D-(-)-Leucinol ≥95%
D-(-)-Leucinol

Hazard: Irritant

Formula: C₆H₁₅NO
MW: 117.19 g/mol
MDL Number: MFCD00004734
CAS Number: 53448-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR