2-Thenoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Thiophene-2-carboxylic acid, 2-Thiophenecarboxylic acid

074924-100G 074924-25G
102538-892EA 52.31 USD
102538-892 102551-924
2-Thenoic acid ≥95%
2-Thenoic acid

Hazard: Irritant

Formula: C₅H₄O₂S
MW: 128.15 g/mol
Boiling Pt: 259-261°C
Melting Pt: 126-130°C
MDL Number: MFCD00005437
CAS Number: 527-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR