2,5-Xylylacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2,5-Dimethylphenyl)acetic acid, (2,5-Dimethylphenyl)acetic acid

074923-100G 074923-25G
102538-888EA 375.38 USD
102538-888 102833-740
2,5-Xylylacetic acid ≥95%
2,5-Xylylacetic acid

Hazard: Irritant

Formula: (C₆H₃(CH₃)₂)CH₂COOH
MW: 164.2 g/mol
MDL Number: MFCD00014369
CAS Number: 13612-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR