N-Boc-cyclopropylamine ≥95%

Supplier: Matrix Scientific

Synonyms: Cyclopropylamine, N-BOC protected, N-Boc-Cyclopropylamine

074921-1G 074921-5G 074921-10G
102538-876EA 107.69 USD
102538-876 102538-878 102538-880
N-Boc-cyclopropylamine ≥95%
N-Boc-cyclopropylamine

Hazard: Irritant

Formula: C₈H₁₅NO₂
MW: 157.2102 g/mol
MDL Number: MFCD09701238
CAS Number: 132844-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR