(R)-tert-Butyl(2-oxooxetan-3-yl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: N-Boc-D-serine-β-lactone, (R)-tert-Butyl (2-oxooxetan-3-yl)carbamate

074920-250MG 074920-1G
102538-872EA 541.54 USD
102538-872 102538-874
(R)-tert-Butyl(2-oxooxetan-3-yl)carbamate ≥95%
(R)-tert-Butyl(2-oxooxetan-3-yl)carbamate

Hazard: Irritant

Formula: C₈H₁₃NO₄
MW: 187.1900 g/mol
MDL Number: MFCD11519129
CAS Number: 126330-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR