4-Ethylphenylacetylene ≥95%

Supplier: Matrix Scientific

Synonyms: (4-Ethylphenyl)acetylene, 1-Ethyl-4-ethynylbenzene, 1-Ethynyl-4-ethylbenzene, (4-Ethylphenyl)ethyne

074914-1G 074914-5G 074914-10G
102538-838EA 55.38 USD
102538-838 102538-840 102538-842
4-Ethylphenylacetylene ≥95%
4-Ethylphenylacetylene

Hazard: Irritant

Formula: C₁₀H₁₀
MW: 130.19 g/mol
MDL Number: MFCD00173887
CAS Number: 40307-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR