4-Bromo-3-chloroanisole ≥95%

Supplier: Matrix Scientific

074913-5G 074913-10G 074913-100G
102538-832EA 64.62 USD
102538-832 102538-834 102538-836
4-Bromo-3-chloroanisole ≥95%
4-Bromo-3-chloroanisole

Hazard: Irritant

Formula: C₇H₆BrClO
MW: 221.48 g/mol
MDL Number: MFCD00070742
CAS Number: 50638-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR