Methyl-2-hydroxy-3-phenylpropanoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-hydroxy-3-phenylpropanoate
074911-1G
074911-5G
102538-822EA
427.69
USD
102538-822
102538-824
Methyl-2-hydroxy-3-phenylpropanoate ≥95%
Methyl-2-hydroxy-3-phenylpropanoate
Hazard: Irritant
Formula:
C₁₀H₁₂O₃ MW: 180.21 g/mol |
MDL Number:
MFCD06654233 CAS Number: 13674-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...