(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride ≥95%

Supplier: Matrix Scientific

074910-250MG 074910-1G 074910-5G
102538-816EA 498.46 USD
102538-816 102538-818 102538-820
(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride ≥95%
(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride

Hazard: Irritant

Formula: C₈H₁₀ClF₂N
MW: 193.63 g/mol
MDL Number: MFCD11101244
CAS Number: 844647-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR