Aminoacetone hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Aminopropan-2-one hydrochloride

074908-1G 074908-5G
102538-804EA 89.23 USD
102538-804 102538-806
Aminoacetone hydrochloride ≥95%
Aminoacetone hydrochloride

Hazard: Irritant

Formula: C₃H₈ClNO
MDL Number: MFCD00056125
CAS Number: 7737-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR