4-Fluorophenethyl bromide ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Fluoro-4-(2-bromoethyl)benzene, 1-(2-bromoethyl)-4-fluorobenzene
074904-1G
074904-5G
074904-10G
102538-780EA
33.85
USD
102538-780
102538-782
102538-784
4-Fluorophenethyl bromide ≥95%
4-Fluorophenethyl bromide
Hazard: Irritant
Formula:
C₈H₈BrF MW: 203.05 g/mol |
MDL Number:
MFCD03844794 CAS Number: 332-42-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...