4-Nitrophenethyl bromide ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(4-Nitrophenyl)-1-bromoethane, 1-(2-Bromoethyl)-4-nitrobenzene
074902-100G
074902-25G
102538-774EA
176.92
USD
102538-774
102833-738
4-Nitrophenethyl bromide ≥95%
4-Nitrophenethyl bromide
Hazard: Irritant
Formula:
C₈H₈BrNO₂ MW: 230.06 g/mol |
MDL Number:
MFCD00007386 CAS Number: 5339-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...