H-D-Arg(Pbf)-OH ≥95%

Supplier: Matrix Scientific

074901-1G 074901-5G 074901-10G
102538-764EA 53.85 USD
102538-764 102538-766 102538-768
H-D-Arg(Pbf)-OH ≥95%
H-D-Arg(Pbf)-OH

Hazard: Irritant

Formula: C₁₉H₃₀N₄O₅S
MDL Number: MFCD00236798
CAS Number: 200116-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR