Methyl-1-methyl-1H-[1,2,4]triazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 1-methyl-1H-[1,2,4]triazole-3-carboxylate

074900-250MG 074900-1G
102538-758EA 223.08 USD
102538-758 102538-760
Methyl-1-methyl-1H-[1,2,4]triazole-3-carboxylate ≥95%
Methyl-1-methyl-1H-[1,2,4]triazole-3-carboxylate

Hazard: Irritant

Formula: C₅H₇N₃O₂
MW: 141.1300 g/mol
MDL Number: MFCD13191558
CAS Number: 57031-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR