3',4'-Dichloroacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(3,4-dichlorophenyl)ethanone

074898-25G 074898-100G
102538-750EA 49.23 USD
102538-750 102538-752
3',4'-Dichloroacetophenone ≥95%
3',4'-Dichloroacetophenone

Hazard: Irritant

Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
MDL Number: MFCD00000553
CAS Number: 2642-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR