5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

074897-250MG 074897-1G
102538-746EA 276.92 USD
102538-746 102538-748
5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylic acid ≥95%
5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₈N₂O₂
MDL Number: MFCD11846936
CAS Number: 796729-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR