Dimethyl-2-cyanosuccinate ≥95%

Supplier: Matrix Scientific

074896-5G 074896-100G 074896-25G
102538-740EA 60 USD
102538-740 102538-744 102551-922
Dimethyl-2-cyanosuccinate ≥95%
Dimethyl-2-cyanosuccinate

Hazard: Irritant

Formula: C₇H₉NO₄
CAS Number: 6283-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR