3-Bromo-2-chlorobenzoic acid ≥95%
Supplier: Matrix Scientific
074894-1G
074894-5G
102538-734EA
38.46
USD
102538-734
102538-736
3-Bromo-2-chlorobenzoic acid ≥95%
3-Bromo-2-chlorobenzoic acid
Hazard: Irritant
Formula:
C₇H₄BrClO₂ |
MDL Number:
MFCD01569399 CAS Number: 56961-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...