Ethyl-5-aminovalerate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-aminopentanoate hydrochloride

074893-1G 074893-5G
102538-730EA 315.38 USD
102538-730 102538-732
Ethyl-5-aminovalerate hydrochloride ≥95%
Ethyl-5-aminovalerate hydrochloride

Hazard: Irritant

Formula: C₇H₁₆ClNO₂
MW: 181.66 g/mol
MDL Number: MFCD12546205
CAS Number: 29840-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR