Ala-OBzl TosOH ≥95%

Supplier: Matrix Scientific

Synonyms: L-Alanine Benzyl Ester p-Toluenesulphonate, L-Alanine benzyl ester p-toluenesulphonate salt, salt H-Ala-OBzl.Tos-OH, H-Ala-OBzl·p-tosylate, H-Ala-OBzl.TosOH, H-Ala-OBzl·Tos-OH

074892-5G 074892-10G 074892-100G
102538-724EA 90.77 USD
102538-724 102538-726 102538-728
Ala-OBzl TosOH ≥95%
Ala-OBzl TosOH

Hazard: Irritant

Formula: C₁₇H₂₁NO₅S
MDL Number: MFCD00066143
CAS Number: 42854-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR