(R)-Methyl-2-amino-2-(4-hydroxyphenyl)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-Methyl 2-amino-2-(4-hydroxyphenyl)acetate

074888-1G 074888-5G 074888-10G
102538-702EA 15.38 USD
102538-702 102538-704 102538-706
(R)-Methyl-2-amino-2-(4-hydroxyphenyl)acetate ≥95%
(R)-Methyl-2-amino-2-(4-hydroxyphenyl)acetate

Hazard: Irritant

Formula: C₉H₁₁NO₃
MW: 181.1885 g/mol
MDL Number: MFCD07778088
CAS Number: 37763-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR