Methyl-2-methyl-4-hydroxy-2H-1,2-benzothiazine-3-carboxylate-1,1-dioxide ≥95%

Supplier: Matrix Scientific

074887-1G 074887-5G
102538-698EA 355.38 USD
102538-698 102538-700
Methyl-2-methyl-4-hydroxy-2H-1,2-benzothiazine-3-carboxylate-1,1-dioxide ≥95%
Methyl-2-methyl-4-hydroxy-2H-1,2-benzothiazine-3-carboxylate-1,1-dioxide

Hazard: Irritant

Formula: C₁₁H₁₁NO₅S
MDL Number: MFCD00071766
CAS Number: 35511-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR