(R)-(-)-2-Aminobutyric acid ≥95%

Supplier: Matrix Scientific

074881-10G 074881-100G
102538-668EA 213.85 USD
102538-668 102538-670
(R)-(-)-2-Aminobutyric acid ≥95%
(R)-(-)-2-Aminobutyric acid

Hazard: Irritant

Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD00064414
CAS Number: 2623-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR