3-Cyanobenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Formylbenzonitrile

074878-5G 074878-10G
102538-654EA 30.77 USD
102538-654 102538-656
3-Cyanobenzaldehyde ≥95%
3-Cyanobenzaldehyde

Hazard: Irritant

Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 210
MDL Number: MFCD00003344
CAS Number: 24964-64-5
UN: 2811
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR