4-(1-Pyrrolidinyl)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Pyrrolidinopyridine

074876-10G 074876-100G
102538-646EA 47.69 USD
102538-646 102538-648
4-(1-Pyrrolidinyl)pyridine ≥95%
4-(1-Pyrrolidinyl)pyridine

Hazard: Irritant

Formula: C₉H₁₂N₂
MW: 148.21 g/mol
MDL Number: MFCD00006415
CAS Number: 2456-81-7
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR