(S)-(-)-ɑ-Amino-γ-butyrolactone hydrochloride ≥95%
Supplier: Matrix Scientific
074872-1G
074872-5G
102538-628EA
100
USD
102538-628
102538-630
(S)-(-)-ɑ-Amino-γ-butyrolactone hydrochloride ≥95%
(S)-(-)-alpha-Amino-gamma-butyrolactone hydrochloride
Hazard: Irritant
Formula:
C₄H₇NO₂ MW: 137.56 g/mol Melting Pt: 219 °C |
MDL Number:
MFCD00058172 CAS Number: 2185-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...