D-Valine methyl ester hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: H-D-Val-OMe.HCl, H-D-Val-OMe·HCl

074871-10G 074871-100G
102538-624EA 123.08 USD
102538-624 102538-626
D-Valine methyl ester hydrochloride ≥95%
D-Valine methyl ester hydrochloride

Hazard: Irritant

Formula: C₆H₁₃NO₂
MW: 167.633 g/mol
MDL Number: MFCD00237309
CAS Number: 7146-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR