(S)-(-)-N-Benzyl-α-methylbenzylamine ≥95%

Supplier: Matrix Scientific

074870-100G 074870-25G
102538-620EA 286.15 USD
102538-620 102833-732
(S)-(-)-N-Benzyl-α-methylbenzylamine ≥95%
(S)-(-)-N-Benzyl-α-methylbenzylamine

Hazard: Irritant

Formula: C₁₅H₁₇N
MDL Number: MFCD00066325
CAS Number: 17480-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR